Please login to your online account to display your discounted pricing

3-Nitrobenzenesulfonic acid, sodium salt, 99%, ACROS Organics™


Quantity 1kg
Packaging Plastic Bottle

Manufacturer: thermo fisher scientific  209120010

 View more versions of this product

Catalog No. AC209120010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 225.15
Quantity 1kg
Color Light yellow
Beilstein 11, 68
Packaging Plastic Bottle
pH 6 to 10 (1% aq. soln.)
CAS Max % 100.0
Molecular Formula C6H4NNaO5S
Linear Formula O2NC6H4SO3Na
Assay Percent Range 99%
Chemical Name or Material 3-Nitrobenzenesulfonic acid, sodium salt
CAS 127-68-4
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
CAS Min % 98.5
Infrared Spectrum Authentic
MDL Number MFCD00007490
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Nitrobenzenesulfonic acid, sodium salt
CAS 127-68-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H4NNaO5S
Linear Formula O2NC6H4SO3Na
Formula Weight 225.15
MDL Number MFCD00007490
Accessories

GHS P Statement:
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.

SDS & Safety Documents