Please login to your online account to display your discounted pricing

3-Nitrobenzenesulfonic acid monohydrate, 95%, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  415810050

 View more versions of this product

Catalog No. AC415810050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 221.2
Physical Form Crystalline Powder
Quantity 5g
Color Light yellow
TSCA TSCA
RTECS Number SZ9430100
EINECS Number 226-261-2
Melting Point 90° to 95°C
CAS Max % 100.0
CAS Min % 94.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 1 Warning
CAS 98-47-5
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
MDL Number MFCD00151046
Chemical Name or Material 3-Nitrobenzenesulfonic acid monohydrate
Molecular Formula C6H5NO5S·H2O
CAS 79326-96-8
Name Note 95%

Specifications

Chemical Name or Material 3-Nitrobenzenesulfonic acid monohydrate
Name Note 95%
CAS 79326-96-8
CAS Min % 94.0
CAS Max % 100.0
CAS 98-47-5
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H5NO5S·H2O
Formula Weight 221.2
MDL Number MFCD00151046
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents