missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrobenzenesulfonamide, 99%, Thermo Scientific™

$55.23 - $189.19

Chemical Identifiers

CAS 121-52-8
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.184
MDL Number MFCD00007935
InChI Key TXTQURPQLVHJRE-UHFFFAOYSA-N
Synonym benzenesulfonamide, 3-nitro, m-nitrobenzenesulfonamide, 3-nitrobenzolesulfamide, 3-nitrobenzenesulphonamide, 3-nitrobenzene-1-sulfonamide, m-nitrobenzenesulphonamide, 3-nitro-benzenesulfonamide, benzenesulfonamide, m-nitro, 3-nitrobenzene sulfonamide, m-nitrobenzolsulfonamid
PubChem CID 67138
IUPAC Name 3-nitrobenzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5583103
View Documents Promotion Details
Thermo Scientific Chemicals
H5583103
1 g
Each for $55.23
 
AAH5583106
View Documents Promotion Details
Thermo Scientific Chemicals
H5583106
5 g
Each for $189.19
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

121-52-8
202.184
TXTQURPQLVHJRE-UHFFFAOYSA-N
67138
C1=CC(=CC(=C1)S(=O)(=O)N)[N+](=O)[O-]
C6H6N2O4S
MFCD00007935
benzenesulfonamide, 3-nitro, m-nitrobenzenesulfonamide, 3-nitrobenzolesulfamide, 3-nitrobenzenesulphonamide, 3-nitrobenzene-1-sulfonamide, m-nitrobenzenesulphonamide, 3-nitro-benzenesulfonamide, benzenesulfonamide, m-nitro, 3-nitrobenzene sulfonamide, m-nitrobenzolsulfonamid
3-nitrobenzenesulfonamide

Specifications

121-52-8
White
MFCD00007935
benzenesulfonamide, 3-nitro, m-nitrobenzenesulfonamide, 3-nitrobenzolesulfamide, 3-nitrobenzenesulphonamide, 3-nitrobenzene-1-sulfonamide, m-nitrobenzenesulphonamide, 3-nitro-benzenesulfonamide, benzenesulfonamide, m-nitro, 3-nitrobenzene sulfonamide, m-nitrobenzolsulfonamid
C1=CC(=CC(=C1)S(=O)(=O)N)[N+](=O)[O-]
202.184
202.19
Powder
166°C to 168°C
C6H6N2O4S
1 g
TXTQURPQLVHJRE-UHFFFAOYSA-N
3-nitrobenzenesulfonamide
67138
99%
3-Nitrobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 204-477-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'