missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitrobenzenesulfonamide, 99%

Manufacturer:  Alfa Aesar™ H5583114

 View more versions of this product

Catalog No. AAH5583114



Specifications

Specifications

3-Nitrobenzenesulfonamide
166°C to 168°C
121-52-8
MFCD00007935
TXTQURPQLVHJRE-UHFFFAOYSA-N
3-nitrobenzenesulfonamide
67138
Powder
White
25g
C6H6N2O4S
benzenesulfonamide, 3-nitro, m-nitrobenzenesulfonamide, 3-nitrobenzolesulfamide, 3-nitrobenzenesulphonamide, 3-nitrobenzene-1-sulfonamide, m-nitrobenzenesulphonamide, 3-nitro-benzenesulfonamide, benzenesulfonamide, m-nitro, 3-nitrobenzene sulfonamide, m-nitrobenzolsulfonamid
C1=CC(=CC(=C1)S(=O)(=O)N)[N+](=O)[O-]
202.184
202.19
99%

Chemical Identifiers

121-52-8
202.184
TXTQURPQLVHJRE-UHFFFAOYSA-N
67138
C1=CC(=CC(=C1)S(=O)(=O)N)[N+](=O)[O-]
C6H6N2O4S
MFCD00007935
benzenesulfonamide, 3-nitro, m-nitrobenzenesulfonamide, 3-nitrobenzolesulfamide, 3-nitrobenzenesulphonamide, 3-nitrobenzene-1-sulfonamide, m-nitrobenzenesulphonamide, 3-nitro-benzenesulfonamide, benzenesulfonamide, m-nitro, 3-nitrobenzene sulfonamide, m-nitrobenzolsulfonamid
3-nitrobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents