Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitrobenzeneboronic acid L-tartaric acid ester, 1M soln. in THF

Chemical Identifiers

Chemical Name or Material 3-Nitrobenzeneboronic acid L-tartaric acid ester
CAS 467443-01-2
Formula Weight 280.99
MDL Number MFCD09750458
UN Number UN2056
Synonym 3-Nitrophenylboronic acid L-tartaric acid ester, 1M soln. in THF
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH55216AA

 
alfa aesar™
H55216AA
5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55216AC

 
alfa aesar™
H55216AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 280.99
Chemical Name or Material 3-Nitrobenzeneboronic acid L-tartaric acid ester
Physical Form Liquid
Refractive Index 1.4508
MDL Number MFCD09750458
CAS 467443-01-2
UN Number UN2056
Synonym 3-Nitrophenylboronic acid L-tartaric acid ester, 1M soln. in THF
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: TETRAHYDROFURAN
Concentration 1M soln. in THF
Density 1.053g/mL
Sensitivity Moisture Sensitive
Color Colorless to brown
Quantity 5mL
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.