missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Nitrobenzeneboronic acid, 98%, Thermo Scientific Chemicals
$42.38 - $381.89
Chemical Identifiers
CAS | 13331-27-6 |
---|---|
Molecular Formula | C6H6BNO4 |
Molecular Weight (g/mol) | 166.93 |
MDL Number | MFCD00007193 |
InChI Key | ZNRGSYUVFVNSAW-UHFFFAOYSA-N |
Synonym | 3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol |
PubChem CID | 1677 |
IUPAC Name | (3-nitrophenyl)boronic acid |
SMILES | OB(O)C1=CC=CC(=C1)[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1333603
|
Thermo Scientific Chemicals
A1333603 |
1 g |
Each for $42.38
|
|
|||||
AAA1333606
|
Thermo Scientific Chemicals
A1333606 |
5 g |
Each for $108.48
|
|
|||||
AAA1333614
|
Thermo Scientific Chemicals
A1333614 |
25 g |
Each for $381.89
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13331-27-6 | |
166.93 | |
ZNRGSYUVFVNSAW-UHFFFAOYSA-N | |
1677 | |
OB(O)C1=CC=CC(=C1)[N+]([O-])=O |
C6H6BNO4 | |
MFCD00007193 | |
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol | |
(3-nitrophenyl)boronic acid |
Specifications
13331-27-6 | |
C6H6BNO4 | |
1 g | |
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol | |
OB(O)C1=CC=CC(=C1)[N+]([O-])=O | |
166.93 | |
166.93 | |
3-Nitrobenzeneboronic acid |
∼285°C (decomposition) | |
MFCD00007193 | |
2938638 | |
ZNRGSYUVFVNSAW-UHFFFAOYSA-N | |
(3-nitrophenyl)boronic acid | |
1677 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : CY8980000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only