Learn More
3-Nitrobenzamide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1548130
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Nitrobenzamide | |
140°C to 143°C | |
MFCD00007984 | |
777185 | |
KWAYEPXDGHYGRW-UHFFFAOYSA-N | |
3-nitrobenzamide | |
12576 | |
98% |
645-09-0 | |
C7H6N2O3 | |
250 g | |
m-nitrobenzamide, benzamide, 3-nitro, benzamide, m-nitro, 3-nitro-benzamide, 3-nitrophenylcarboxamide, acmc-209nmr, 3-nitrobenzamide, dsstox_cid_5732, dsstox_rid_77897, dsstox_gsid_25732 | |
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)N | |
166.136 | |
166.14 |
Chemical Identifiers
645-09-0 | |
166.136 | |
KWAYEPXDGHYGRW-UHFFFAOYSA-N | |
12576 | |
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)N |
C7H6N2O3 | |
MFCD00007984 | |
m-nitrobenzamide, benzamide, 3-nitro, benzamide, m-nitro, 3-nitro-benzamide, 3-nitrophenylcarboxamide, acmc-209nmr, 3-nitrobenzamide, dsstox_cid_5732, dsstox_rid_77897, dsstox_gsid_25732 | |
3-nitrobenzamide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 211-431-0
RTECSNumber : CV5601250
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only