Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitrobenzaldoxime, 98%

Chemical Identifiers

Chemical Name or Material 3-Nitrobenzaldoxime
CAS 3431-62-7
Molecular Formula C7H6N2O3
Formula Weight 166.14
MDL Number MFCD00007275
UN Number UN2811
Beilstein 2047081
Synonym 3-Nitrobenzaldehyde oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0914414

  SDS
alfa aesar™
L0914414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0914422

  SDS
alfa aesar™
L0914422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.14
CAS 3431-62-7
Chemical Name or Material 3-Nitrobenzaldoxime
Molecular Formula C7H6N2O3
MDL Number MFCD00007275
UN Number UN2811
Synonym 3-Nitrobenzaldehyde oxime
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 122° to 125°C
Assay 98%
Beilstein 2047081
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.