Learn More
3-Nitroaniline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 128361000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Nitroaniline | |
99-09-2 | |
100.0 | |
Orange to Yellow | |
196°C | |
97.5% min. (GC) | |
C6H6N2O2 | |
MFCD00007782 | |
12, 698 | |
m-nitroaniline, benzenamine, 3-nitro, 3-nitrophenylamine, nitranilin, 3-aminonitrobenzene, fast orange base r, azobase mna, devol orange r, m-nitrophenylamine, 3-nitrobenzenamine | |
XJCVRTZCHMZPBD-UHFFFAOYSA-N | |
3-nitroaniline | |
7423 | |
98% |
99+% | |
97.5 | |
111.0°C to 114.0°C | |
306.0°C | |
Authentic | |
Glass bottle | |
O2NC6H4NH2 | |
100 g | |
15, 6668 | |
Solubility in water: 1.25g/L in water. Other solubilities: soluble in alcohol,ether,methanol and ethanol,1 g soluble in 20 mL alcohol,1 g soluble in 18 mL ether,1 g soluble in 11.5 mL methanol | |
NC1=CC=CC(=C1)[N+]([O-])=O | |
138.13 | |
138.13 | |
Crystalline Powder, Crystals or Chunks |
Chemical Identifiers
99-09-2 | |
138.13 | |
XJCVRTZCHMZPBD-UHFFFAOYSA-N | |
7423 | |
NC1=CC=CC(=C1)[N+]([O-])=O |
C6H6N2O2 | |
MFCD00007782 | |
m-nitroaniline, benzenamine, 3-nitro, 3-nitrophenylamine, nitranilin, 3-aminonitrobenzene, fast orange base r, azobase mna, devol orange r, m-nitrophenylamine, 3-nitrobenzenamine | |
3-nitroaniline |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Harmful to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
GHS Signal Word: Danger
EINECSNumber : 202-729-1
RTECSNumber : BY6825000
TSCA : TSCA
RUO – Research Use Only