missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3'-Nitroacetophenone, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1215530
Description
3'-Nitroacetophenone is used as an intermediate in pharmaceutical industry for making drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3′-Nitroacetophenone is used as an intermediate in pharmaceutical industry for making drugs.
Solubility
Soluble in water 0.75 g/L @ 20°C.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Specifications
3'-Nitroacetophenone | |
76°C to 80°C | |
Slightly sweetish | |
MFCD00007259 | |
743002 | |
Soluble in water 0.75g/L @ 20°C. | |
CC(=O)C1=CC(=CC=C1)[N+](=O)[O-] | |
165.148 | |
165.15 |
121-89-1 | |
202°C | |
C8H7NO3 | |
250 g | |
3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon | |
ARKIFHPFTHVKDT-UHFFFAOYSA-N | |
1-(3-nitrophenyl)ethanone | |
8494 | |
≥98% |
Chemical Identifiers
121-89-1 | |
165.148 | |
ARKIFHPFTHVKDT-UHFFFAOYSA-N | |
8494 | |
CC(=O)C1=CC(=CC=C1)[N+](=O)[O-] |
C8H7NO3 | |
MFCD00007259 | |
3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon | |
1-(3-nitrophenyl)ethanone |
Safety and Handling
EINECSNumber : 204-504-3
RTECSNumber : AM9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only