Please login to your online account to display your discounted pricing

3'-Nitroacetanilide 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3';-Nitroacetanilide
CAS 122-28-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H8N2O3
Linear Formula CH3C(O)NHC6H4NO2
Formula Weight 180.16
MDL Number MFCD00017015
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC387470050

  SDS
thermo fisher scientific
AC387470050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 180.16
Physical Form Crystalline To Powder
Quantity 5g
Color Beige
Melting Point 150° to 154°C
Merck Index 15, 6667
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula CH3C(O)NHC6H4NO2
CAS 122-28-1
Molecular Formula C8H8N2O3
Chemical Name or Material 3';-Nitroacetanilide
Assay Percent Range 99%
MDL Number MFCD00017015
CAS Min % 98.5
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.