missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Nitroacetanilide, 98+%

Manufacturer:  Alfa Aesar™ B2206222

 View more versions of this product

Catalog No. AAB2206222



Description

Description

Specifications

Specifications

3'-Nitroacetanilide
100g
C8H8N2O3
2211961
3'-nitroacetanilide, 3-nitro-n-acetylaniline, 3-nitroacetanilide, acetamide, n-3-nitrophenyl, acetanilide, 3'-nitro, m-nitroacetanilide, n-3-nitrophenyl acetamide, n-acetyl-m-nitroaniline, qgh8s22nbp, unii-qgh8s22nbp
CC(=O)NC1=CC(=CC=C1)[N+](=O)[O-]
180.163
180.16
151°C to 153°C
122-28-1
MFCD00017015
14,6581
KFTYNYHJHKCRKU-UHFFFAOYSA-N
N-(3-nitrophenyl)acetamide
31206
≥98%

Chemical Identifiers

122-28-1
180.163
KFTYNYHJHKCRKU-UHFFFAOYSA-N
31206
CC(=O)NC1=CC(=CC=C1)[N+](=O)[O-]
C8H8N2O3
MFCD00017015
3'-nitroacetanilide, 3-nitro-n-acetylaniline, 3-nitroacetanilide, acetamide, n-3-nitrophenyl, acetanilide, 3'-nitro, m-nitroacetanilide, n-3-nitrophenyl acetamide, n-acetyl-m-nitroaniline, qgh8s22nbp, unii-qgh8s22nbp
N-(3-nitrophenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 204-532-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents
Product Certifications