missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-o-phenylenediamine, 98%

CAS: 3694-52-8 | C6H7N3O2 | 153.141 g/mol

Supplier:  Thermo Scientific Chemicals B2020114

 View more versions of this product

Catalog No. AAB2020114


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Nitro-o-phenylenediamine
157°C to 162°C
MFCD00007722
909344
IOCXBXZBNOYTLQ-UHFFFAOYSA-N
3-nitrobenzene-1,2-diamine
4359525
98%
3694-52-8
C6H7N3O2
25 g
3-nitro-1,2-phenylenediamine, 3-nitro-o-phenylenediamine, 1,2-diamino-3-nitrobenzene, 3-nitrophenylenediamine, 2-amino-6-nitroaniline, 1,2-benzenediamine, 3-nitro, 3-nitro-1,2-benzenediamine, o-phenylenediamine, 3-nitro, unii-wrz63l50zk, ccris 5427
C1=CC(=C(C(=C1)[N+](=O)[O-])N)N
153.141
153.14

Chemical Identifiers

3694-52-8
153.141
IOCXBXZBNOYTLQ-UHFFFAOYSA-N
4359525
C1=CC(=C(C(=C1)[N+](=O)[O-])N)N
C6H7N3O2
MFCD00007722
3-nitro-1,2-phenylenediamine, 3-nitro-o-phenylenediamine, 1,2-diamino-3-nitrobenzene, 3-nitrophenylenediamine, 2-amino-6-nitroaniline, 1,2-benzenediamine, 3-nitro, 3-nitro-1,2-benzenediamine, o-phenylenediamine, 3-nitro, unii-wrz63l50zk, ccris 5427
3-nitrobenzene-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 223-013-5

RTECSNumber : ST2974000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only