missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitro-N-phenylbenzamide, 97%

$220.53 - $703.20

Chemical Identifiers

CAS 2243-73-4
Molecular Formula C13H10N2O3
Molecular Weight (g/mol) 242.234
MDL Number MFCD00017021
InChI Key VWIRWLAPFZXYSL-UHFFFAOYSA-N
Synonym 3-nitrobenzanilide, 3-nitro-n-phenyl-benzamide, benzamide, 3-nitro-n-phenyl, 3-nitrophenyl-n-benzamide, 3-nitro-n-phenyl benzamide
PubChem CID 347755
IUPAC Name 3-nitro-N-phenylbenzamide
SMILES C1=CC=C(C=C1)NC(=O)C2=CC(=CC=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60695MD View Documents Alfa Aesar™
H60695MD
250mg Each for $220.53
 
AAH6069503 View Documents Alfa Aesar™
H6069503
1g Each for $703.20
 
Specifications

Chemical Identifiers

2243-73-4
242.234
VWIRWLAPFZXYSL-UHFFFAOYSA-N
347755
C1=CC=C(C=C1)NC(=O)C2=CC(=CC=C2)[N+](=O)[O-]
C13H10N2O3
MFCD00017021
3-nitrobenzanilide, 3-nitro-n-phenyl-benzamide, benzamide, 3-nitro-n-phenyl, 3-nitrophenyl-n-benzamide, 3-nitro-n-phenyl benzamide
3-nitro-N-phenylbenzamide

Specifications

250mg
C13H10N2O3
3-nitrobenzanilide, 3-nitro-n-phenyl-benzamide, benzamide, 3-nitro-n-phenyl, 3-nitrophenyl-n-benzamide, 3-nitro-n-phenyl benzamide
C1=CC=C(C=C1)NC(=O)C2=CC(=CC=C2)[N+](=O)[O-]
242.234
242.23
3-Nitro-N-phenylbenzamide
2243-73-4
MFCD00017021
VWIRWLAPFZXYSL-UHFFFAOYSA-N
3-nitro-N-phenylbenzamide
347755
97%