Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitro-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material 3-Nitro-D-phenylalanine
CAS 169530-97-6
Formula Weight 210.19
MDL Number MFCD04117829
Synonym H-D-Phe(3-NO2)-OH; (R)-2-Amino-3-(3-nitrophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6348303

  SDS
alfa aesar™
H6348303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63483MD

  SDS
alfa aesar™
H63483MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.19
CAS 169530-97-6
MDL Number MFCD04117829
Physical Form Solid
Synonym H-D-Phe(3-NO2)-OH; (R)-2-Amino-3-(3-nitrophenyl)propionic acid
Assay 95%
Quantity 1g
Storage Note 1 -30° to -10°C
Color Off-white to pale yellow
Chemical Name or Material 3-Nitro-D-phenylalanine
Safety and Handling

-30° to -10°C