missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Nitro-D-phenylalanine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H63483MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Nitro-D-phenylalanine | |
White to Yellow | |
MFCD04117829 | |
YTHDRUZHNYKZGF-UHFFFAOYNA-N | |
2-amino-3-(3-nitrophenyl)propanoic acid | |
7000156 | |
95% | |
Solid |
169530-97-6 | |
C9H10N2O4 | |
r-2-amino-3-3-nitrophenyl propanoic acid, 3-nitro-d-phenylalanine, d-3-nitrophenylalanine, 2r-2-amino-3-3-nitrophenyl propanoic acid, d-3-no2-phe-oh, d-3-nitrophe, d-3-nitro phenylalanine, r-2-amino-3-3-nitrophenyl propanoicacid | |
NC(CC1=CC(=CC=C1)[N+]([O-])=O)C(O)=O | |
210.19 | |
210.19 | |
250 mg |
Chemical Identifiers
169530-97-6 | |
210.19 | |
YTHDRUZHNYKZGF-UHFFFAOYNA-N | |
7000156 | |
NC(CC1=CC(=CC=C1)[N+]([O-])=O)C(O)=O |
C9H10N2O4 | |
MFCD04117829 | |
r-2-amino-3-3-nitrophenyl propanoic acid, 3-nitro-d-phenylalanine, d-3-nitrophenylalanine, 2r-2-amino-3-3-nitrophenyl propanoic acid, d-3-no2-phe-oh, d-3-nitrophe, d-3-nitro phenylalanine, r-2-amino-3-3-nitrophenyl propanoicacid | |
2-amino-3-(3-nitrophenyl)propanoic acid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only