Learn More
3-Nitro-alpha-toluenesulfonyl chloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2333303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilityReacts with water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
Specifications
3-Nitro-alpha-toluenesulfonyl chloride | |
95°C to 102°C | |
MFCD01859935 | |
UN3261 | |
3-nitrophenyl methanesulfonyl chloride, 3-nitrobenzylsulfonyl chloride, 3-nitrophenylmethanesulfonylchloride, benzenemethanesulfonyl chloride, 3-nitro, 3-nitro-alpha-toluenesulfonyl chloride, chloro 3-nitrophenyl methyl sulfone, acmc-1b04d, 3-nitro-phenylmethane-sulfonyl chloride, 3-nitrophenyl methanesulphonyl chloride, 3-nitrobenzenemethanesulfonyl chloride | |
IUYFKLDRPZLAQQ-UHFFFAOYSA-N | |
(3-nitrophenyl)methanesulfonyl chloride | |
13196775 | |
95% |
58032-84-1 | |
C7H6ClNO4S | |
1 g | |
Moisture sensitive | |
Reacts with water. | |
[O-][N+](=O)C1=CC=CC(CS(Cl)(=O)=O)=C1 | |
235.64 | |
235.64 |
Chemical Identifiers
58032-84-1 | |
235.64 | |
IUYFKLDRPZLAQQ-UHFFFAOYSA-N | |
13196775 | |
[O-][N+](=O)C1=CC=CC(CS(Cl)(=O)=O)=C1 |
C7H6ClNO4S | |
MFCD01859935 | |
3-nitrophenyl methanesulfonyl chloride, 3-nitrobenzylsulfonyl chloride, 3-nitrophenylmethanesulfonylchloride, benzenemethanesulfonyl chloride, 3-nitro, 3-nitro-alpha-toluenesulfonyl chloride, chloro 3-nitrophenyl methyl sulfone, acmc-1b04d, 3-nitro-phenylmethane-sulfonyl chloride, 3-nitrophenyl methanesulphonyl chloride, 3-nitrobenzenemethanesulfonyl chloride | |
(3-nitrophenyl)methanesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only