Please login to your online account to display your discounted pricing

3-Nitro-alpha,alpha,alpha-trifluorotoluene, 97%, ACROS Organics™


Quantity 50g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  325700500

 View more versions of this product

Catalog No. AC325700500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 191.11
Quantity 50g
Color Clear yellow
Density 1.4300g/mL
TSCA TSCA
RTECS Number XT3500000
EINECS Number 202-670-1
Beilstein 05, 327
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4710 to 1.4730 (20°C, 589nm)
Synonym 3-Nitrobenzotrifluoride
Packaging Plastic bottle
Infrared Spectrum Authentic
Chemical Name or Material 3-Nitrobenzotrifluoride
MDL Number MFCD00007260
Health Hazard 1 GHS Signal Word: Warning
CAS 98-46-4
Name Note 97%
Assay Percent Range 97%
Molecular Formula C7H4F3NO2

Specifications

Chemical Name or Material 3-Nitrobenzotrifluoride
Name Note 97%
CAS 98-46-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4F3NO2
Formula Weight 191.11
MDL Number MFCD00007260
Synonym 3-Nitrobenzotrifluoride
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.

SDS & Safety Documents