Please login to your online account to display your discounted pricing

3-Nitro-5-(trifluoromethyl)pyridin-2-ol, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-Nitro-5-(trifluoromethyl)pyridin-2-ol
CAS 33252-64-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H3F3N2O3
Formula Weight 208.09
MDL Number MFCD00153193
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF04994DA

  SDS
fisher scientific company
RF04994DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.09
Quantity 1g
Assay 97%
CAS Min % 97.0
MDL Number MFCD00153193
Chemical Name or Material 3-Nitro-5-(trifluoromethyl)pyridin-2-ol
CAS 33252-64-1
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Assay Percent Range 97%
Molecular Formula C6H3F3N2O3
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.