missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitro-4-(trifluoromethoxy)aniline, 95%

Manufacturer:  Alfa Aesar™ H6145622

 View more versions of this product

Catalog No. AAH6145622



Specifications

Specifications

3-Nitro-4-(trifluoromethoxy)aniline
1.5250
100g
C7H5F3N2O3
3-nitro-4-trifluoromethoxy aniline, 4-trifluoromethoxy-3-nitroaniline, benzenamine,3-nitro-4-trifluoromethoxy, 3-nitro-4-trifluoromethoxy-phenylamine, 5-amino-2-trifluoromethoxy nitrobenzene, pubchem4499, acmc-1cmet, 3-nitro-4-trifluormethoxy-aniline, 3-nitro-4-trifluoromethyloxy aniline, 3-nitro-4-trifluoromethoxy aniline #
C1=CC(=C(C=C1N)[N+](=O)[O-])OC(F)(F)F
222.123
222.12
95%
1.543g/mL
300°C (760mmHg)
2822-50-6
MFCD03840207
QXIKEFNBFUHDHK-UHFFFAOYSA-N
3-nitro-4-(trifluoromethoxy)aniline
613694
Solid

Chemical Identifiers

2822-50-6
222.123
QXIKEFNBFUHDHK-UHFFFAOYSA-N
613694
C1=CC(=C(C=C1N)[N+](=O)[O-])OC(F)(F)F
C7H5F3N2O3
MFCD03840207
3-nitro-4-trifluoromethoxy aniline, 4-trifluoromethoxy-3-nitroaniline, benzenamine,3-nitro-4-trifluoromethoxy, 3-nitro-4-trifluoromethoxy-phenylamine, 5-amino-2-trifluoromethoxy nitrobenzene, pubchem4499, acmc-1cmet, 3-nitro-4-trifluormethoxy-aniline, 3-nitro-4-trifluoromethyloxy aniline, 3-nitro-4-trifluoromethoxy aniline #
3-nitro-4-(trifluoromethoxy)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

SDS Documents