missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitro-4-quinolinol, TCI America™

$175.70 - $606.55

Chemical Identifiers

CAS 50332-66-6
Molecular Formula C9H6N2O3
Molecular Weight (g/mol) 190.158
MDL Number MFCD00277722
InChI Key ZWISCKSGNCMAQO-UHFFFAOYSA-N
Synonym 4-Hydroxy-3-nitroquinoline
PubChem CID 316988
IUPAC Name 3-nitro-1H-quinolin-4-one
SMILES C1=CC=C2C(=C1)C(=O)C(=CN2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N09921G View Documents TCI America
N09921G
1g Each for $606.55
 
N0992200MG View Documents TCI America
N0992200MG
200mg Each for $175.70
 
Specifications

Chemical Identifiers

50332-66-6
190.158
ZWISCKSGNCMAQO-UHFFFAOYSA-N
316988
C1=CC=C2C(=C1)C(=O)C(=CN2)[N+](=O)[O-]
C9H6N2O3
MFCD00277722
4-Hydroxy-3-nitroquinoline
3-nitro-1H-quinolin-4-one

Specifications

Yellow
1g
C9H6N2O3
4-Hydroxy-3-nitroquinoline
C1=CC=C2C(=C1)C(=O)C(=CN2)[N+](=O)[O-]
190.158
190.16
≥98.0% (GC,T)
361°C
50332-66-6
MFCD00277722
ZWISCKSGNCMAQO-UHFFFAOYSA-N
3-nitro-1H-quinolin-4-one
316988
Crystal-Powder at 20°C
3-Nitro-4-quinolinol
Safety and Handling

Safety and Handling

TSCA : No