Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitro-4-(1-piperidinyl)benzaldehyde, 96%

$68.70 - $235.05

Chemical Identifiers

Chemical Name or Material 3-Nitro-4-(1-piperidinyl)benzaldehyde
CAS 39911-29-0
Molecular Formula C12H14N2O3
Formula Weight 234.26
MDL Number MFCD00831780
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2238103

  SDS
alfa aesar™
B2238103
1g Each for $68.70

AAB2238106

  SDS
alfa aesar™
B2238106
5g Each for $235.05
Description & Specifications

Specifications

Formula Weight 234.26
Quantity 1g
Health Hazard 1 Warning
Melting Point 50° to 54°C
Molecular Formula C12H14N2O3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 39911-29-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
Chemical Name or Material 3-Nitro-4-(1-piperidinyl)benzaldehyde
MDL Number MFCD00831780
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.