missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitro-2,6-lutidine, 99%, ACROS Organics™

Chemical Identifiers

CAS 15513-52-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00023503
InChI Key AETHUDGJSSKZKT-UHFFFAOYSA-N
Synonym 2,6-dimethyl-3-nitro-pyridine
PubChem CID 84958
IUPAC Name 2,6-dimethyl-3-nitropyridine
SMILES CC1=NC(=C(C=C1)[N+](=O)[O-])C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC348150010 SDS View Documents Thermo Fisher Scientific
AC348150010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC348150050 SDS View Documents Thermo Fisher Scientific
AC348150050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

15513-52-7
152.15
AETHUDGJSSKZKT-UHFFFAOYSA-N
84958
CC1=NC(=C(C=C1)[N+](=O)[O-])C
C7H8N2O2
MFCD00023503
2,6-dimethyl-3-nitro-pyridine
2,6-dimethyl-3-nitropyridine

Specifications

37.78°C
Glass bottle
32.0°C to 38.0°C
15513-52-7
100.0
C7H8N2O2
20, V,6, 52
AETHUDGJSSKZKT-UHFFFAOYSA-N
2,6-dimethyl-3-nitropyridine
84958
Powder
3-Nitro-2, 6-lutidine
Authentic
Tan to White
1g
98.5
98.5% min. (HPLC)
MFCD00023503
2,6-dimethyl-3-nitro-pyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])C
152.15
152.15
99%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Flammable solid.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 239-543-5