missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(N-tert-Butoxycarbonyl-N-methylamino)pyrrolidine 96.0+%, TCI America™

$283.68 - $849.41

Chemical Identifiers

CAS 172478-00-1
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD00191665
InChI Key XYKYUXYNQDXZTD-UHFFFAOYSA-N
Synonym 3-(N-Boc-N-methylamino)pyrrolidine
PubChem CID 2761404
IUPAC Name tert-butyl N-methyl-N-pyrrolidin-3-ylcarbamate
SMILES CC(C)(C)OC(=O)N(C)C1CCNC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M10965G
View Documents Promotion Details
TCI America
M10965G
5 g
Each for $283.68
 
M109625G
View Documents Promotion Details
TCI America
M109625G
25 g
Each for $849.41
 
Description

Description

Specifications

Chemical Identifiers

172478-00-1
200.282
XYKYUXYNQDXZTD-UHFFFAOYSA-N
2761404
CC(C)(C)OC(=O)N(C)C1CCNC1
C10H20N2O2
MFCD00191665
3-(N-Boc-N-methylamino)pyrrolidine
tert-butyl N-methyl-N-pyrrolidin-3-ylcarbamate

Specifications

172478-00-1
Colorless
MFCD00191665
XYKYUXYNQDXZTD-UHFFFAOYSA-N
tert-butyl N-methyl-N-pyrrolidin-3-ylcarbamate
2761404
≥96.0% (GC)
Liquid
96°C
C10H20N2O2
3-(N-Boc-N-methylamino)pyrrolidine
CC(C)(C)OC(=O)N(C)C1CCNC1
200.282
200.28
5 g
3-(N-tert-Butoxycarbonyl-N-methylamino)pyrrolidine
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'