Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-n-Propylidenephthalide, (E)+(Z), 96%

$20.78 - $629.82

Chemical Identifiers

Chemical Name or Material 3-n-Propylidenephthalide
CAS 17369-59-4
Name Note (E)+(Z)
Molecular Formula C11H10O2
Formula Weight 174.2
MDL Number MFCD00051813
Beilstein 4486
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1331914

  SDS
alfa aesar™
A1331914
25g Each for $65.69

AAA1331906

  SDS
alfa aesar™
A1331906
5g Each for $20.78

AAA1331936

  SDS
alfa aesar™
A1331936
500g Each for $629.82

AAA1331922

  SDS
alfa aesar™
A1331922
100g Each for $194.38
Description & Specifications

Specifications

Formula Weight 174.2
Chemical Name or Material 3-n-Propylidenephthalide
TSCA TSCA
Name Note (E)+(Z)
Assay 96%
Molecular Formula C11H10O2
MDL Number MFCD00051813
CAS 17369-59-4
Beilstein 4486
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number NP7235000
Flash Point 144°C (291°F)
Boiling Point 101° to 103°C (0.01mmHg)
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.