missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-n-Octylthiophene, 97%

Manufacturer:  Alfa Aesar™ H5546614

 View more versions of this product

Catalog No. AAH5546614



Description

Description

Applications
3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).

Solubility
Soluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

3-n-Octylthiophene
113°C (235°F)
106°C to 107°C (3mmHg)
25g
C12H20S
3-n-octylthiophene
WQYWXQCOYRZFAV-UHFFFAOYSA-N
3-octylthiophene
566852
Liquid
0.92 g/mL
1.492
Colorless
65016-62-8
MFCD00085281
Soluble in organic solvents.
CCCCCCCCC1=CSC=C1
196.352
196.35
97%

Chemical Identifiers

65016-62-8
196.352
WQYWXQCOYRZFAV-UHFFFAOYSA-N
566852
CCCCCCCCC1=CSC=C1
C12H20S
MFCD00085281
3-n-octylthiophene
3-octylthiophene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

Documents