Learn More
3-n-Octylthiophene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5546606
Description
3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).
Solubility
Soluble in organic solvents.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
3-n-Octylthiophene | |
Colorless | |
106°C to 107°C (3 mmHg) | |
C12H20S | |
MFCD00085281 | |
3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691 | |
WQYWXQCOYRZFAV-UHFFFAOYSA-N | |
3-octylthiophene | |
566852 | |
97% |
65016-62-8 | |
0.92 g/mL | |
113°C (235°F) | |
1.492 | |
5 g | |
Soluble in organic solvents. | |
CCCCCCCCC1=CSC=C1 | |
196.352 | |
196.35 | |
Liquid |
Chemical Identifiers
65016-62-8 | |
196.352 | |
WQYWXQCOYRZFAV-UHFFFAOYSA-N | |
566852 | |
CCCCCCCCC1=CSC=C1 |
C12H20S | |
MFCD00085281 | |
3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691 | |
3-octylthiophene |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only