missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester, 97+%

$43.24 - $130.38

Chemical Identifiers

CAS 957061-17-5
Molecular Formula C15H22BNO4
Formula Weight 291.16
MDL Number MFCD09258775
Synonym [3-(N,O-Dimethyl hydroxylaminocarbonyl)phenyl] boronic acid pinacol ester; N,O-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzohydroxamate
Chemical Name or Material 3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2794903 SDS Alfa Aesar™
H2794903
1g Each for $43.24
AAH2794906 SDS Alfa Aesar™
H2794906
5g Each for $130.38
Specifications Description & Specifications

Chemical Identifiers

957061-17-5
291.16
[3-(N,O-Dimethyl hydroxylaminocarbonyl)phenyl] boronic acid pinacol ester; N,O-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzohydroxamate
C15H22BNO4
MFCD09258775
3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester

Specifications

957061-17-5
1g
291.16
[3-(N,O-Dimethyl hydroxylaminocarbonyl)phenyl] boronic acid pinacol ester; N,O-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzohydroxamate
≥97%
69° to 72°C
C15H22BNO4
MFCD09258775
3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester