missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[N-Methyl-N-(2-nitrophenyl)sulfamoyl]benzeneboronic acid, 97%, Alfa Aesar™

$226.85 - $725.91

Chemical Identifiers

CAS 1449133-23-6
Molecular Formula C13H13BN2O6S
Molecular Weight (g/mol) 336.125
MDL Number MFCD20265204
InChI Key DXKDWPNDSRHLBL-UHFFFAOYSA-N
Synonym 3-methyl 2-nitrophenyl sulfamoyl phenylboronic acid
PubChem CID 73995688
IUPAC Name [3-[methyl-(2-nitrophenyl)sulfamoyl]phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)S(=O)(=O)N(C)C2=CC=CC=C2[N+](=O)[O-])(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54208MD View Documents Thermo Fisher Scientific
H54208MD
250mg Each for $226.85
 
AAH5420803 View Documents Thermo Fisher Scientific
H5420803
1g Each for $725.91
 
Specifications

Chemical Identifiers

1449133-23-6
336.125
DXKDWPNDSRHLBL-UHFFFAOYSA-N
73995688
B(C1=CC(=CC=C1)S(=O)(=O)N(C)C2=CC=CC=C2[N+](=O)[O-])(O)O
C13H13BN2O6S
MFCD20265204
3-methyl 2-nitrophenyl sulfamoyl phenylboronic acid
[3-[methyl-(2-nitrophenyl)sulfamoyl]phenyl]boronic acid

Specifications

250mg
C13H13BN2O6S
3-methyl 2-nitrophenyl sulfamoyl phenylboronic acid
B(C1=CC(=CC=C1)S(=O)(=O)N(C)C2=CC=CC=C2[N+](=O)[O-])(O)O
336.125
336.13
3-[N-Methyl-N-(2-nitrophenyl)sulfamoyl]benzeneboronic acid
1449133-23-6
MFCD20265204
DXKDWPNDSRHLBL-UHFFFAOYSA-N
[3-[methyl-(2-nitrophenyl)sulfamoyl]phenyl]boronic acid
73995688
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures