Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-n-Hexylpyridine, 98+%

$209.45 - $209.45

Chemical Identifiers

Chemical Name or Material 3-n-Hexylpyridine
CAS 6311-92-8
Molecular Formula C11H17N
Formula Weight 163.27
MDL Number MFCD02683071
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2177509

  SDS
alfa aesar™
B2177509
10g Each for $209.45
Description & Specifications

Specifications

Formula Weight 163.27
MDL Number MFCD02683071
CAS 6311-92-8
Health Hazard 1 Warning
Chemical Name or Material 3-n-Hexylpyridine
Assay ≥98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 10g
Molecular Formula C11H17N
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.