missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-n-Dodecylthiophene, 97%

$71.19 - $813.15

Chemical Identifiers

CAS 104934-52-3
Molecular Formula C16H28S
Molecular Weight (g/mol) 252.46
MDL Number MFCD00130121
InChI Key RFKWIEFTBMACPZ-UHFFFAOYSA-N
Synonym 1-thien-3-yl dodecane
PubChem CID 2733655
IUPAC Name 3-dodecylthiophene
SMILES CCCCCCCCCCCCC1=CSC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5579903 View Documents Alfa Aesar™
H5579903
1g Each for $71.19
 
AAH5579906 View Documents Alfa Aesar™
H5579906
5g Each for $247.78
 
AAH5579914 View Documents Alfa Aesar™
H5579914
25g Each for $813.15
 
Specifications

Chemical Identifiers

104934-52-3
252.46
RFKWIEFTBMACPZ-UHFFFAOYSA-N
2733655
CCCCCCCCCCCCC1=CSC=C1
C16H28S
MFCD00130121
1-thien-3-yl dodecane
3-dodecylthiophene

Specifications

0.902
290°C
104934-52-3
MFCD00130121
RFKWIEFTBMACPZ-UHFFFAOYSA-N
3-dodecylthiophene
2733655
97%
1.488
1g
C16H28S
1-thien-3-yl dodecane
CCCCCCCCCCCCC1=CSC=C1
252.46
252.46
3-n-Dodecylthiophene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures