missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-n-Dodecylthiophene, 97%

Manufacturer:  Alfa Aesar™ H5579903

 View more versions of this product

Catalog No. AAH5579903



Specifications

Specifications

3-n-Dodecylthiophene
1.488
1g
C16H28S
1-thien-3-yl dodecane
CCCCCCCCCCCCC1=CSC=C1
252.46
252.46
0.902
290°C
104934-52-3
MFCD00130121
RFKWIEFTBMACPZ-UHFFFAOYSA-N
3-dodecylthiophene
2733655
97%

Chemical Identifiers

104934-52-3
252.46
RFKWIEFTBMACPZ-UHFFFAOYSA-N
2733655
CCCCCCCCCCCCC1=CSC=C1
C16H28S
MFCD00130121
1-thien-3-yl dodecane
3-dodecylthiophene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents