missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-n-Decylthiophene, 97%

Manufacturer:  Alfa Aesar™ H5525106

 View more versions of this product

Catalog No. AAH5525106



Specifications

Specifications

3-n-Decylthiophene
>113°C (235°F)
119°C to 120°C (1.2mmHg)
65016-55-9
MFCD00143180
JAYBIBLZTQMCAY-UHFFFAOYSA-N
3-decylthiophene
566842
97%
0.912
1.489
5g
C14H24S
3-decylthiophene
CCCCCCCCCCC1=CSC=C1
224.406
224.4

Chemical Identifiers

65016-55-9
224.406
JAYBIBLZTQMCAY-UHFFFAOYSA-N
566842
CCCCCCCCCCC1=CSC=C1
C14H24S
MFCD00143180
3-decylthiophene
3-decylthiophene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents