missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-n-Butyl-2,4-pentanedione, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1123609

 View more versions of this product

Catalog No. AAA1123609



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-n-Butyl-2,4-pentanedione
0.927
C9H16O2
MFCD00026235
507436
MBXOOYPCIDHXGH-UHFFFAOYSA-N
3-butylpentane-2,4-dione
73763
98%
1540-36-9
101°C to 104°C (10 mmHg)
1.447
10 g
3-n-butyl-2,4-pentanedione, 3-butyl-2,4-pentanedione, 3-acetyl-2-heptanone, 3-butylacetylacetone, 2,4-pentanedione, 3-butyl, 3-n-butyl-acetylaceton, 2, 3-butyl, 3-butylpentanedione-2,4, 3-butyl-2,4-pentandione, acmc-209da7
CCCCC(C(C)=O)C(C)=O
156.23
156.23

Chemical Identifiers

1540-36-9
156.23
MBXOOYPCIDHXGH-UHFFFAOYSA-N
73763
CCCCC(C(C)=O)C(C)=O
C9H16O2
MFCD00026235
3-n-butyl-2,4-pentanedione, 3-butyl-2,4-pentanedione, 3-acetyl-2-heptanone, 3-butylacetylacetone, 2,4-pentanedione, 3-butyl, 3-n-butyl-acetylaceton, 2, 3-butyl, 3-butylpentanedione-2,4, 3-butyl-2,4-pentandione, acmc-209da7
3-butylpentane-2,4-dione
Safety and Handling

Safety and Handling

EINECSNumber : 216-274-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only