missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-n-Butyl-2,4-pentanedione, 98%

Manufacturer:  Alfa Aesar™ A1123630

 View more versions of this product

Catalog No. AAA1123630


 

Specifications

Specifications

3-n-Butyl-2,4-pentanedione
98%
MFCD00026235
101°C to 104°C (10mmHg)
3-Acetyl-2-heptanone
CCCCC(C(=O)C)C(=O)C
156.225
156.23
1540-36-9
C9H16O2
507436
250g
MBXOOYPCIDHXGH-UHFFFAOYSA-N
3-butylpentane-2,4-dione
73763

Chemical Identifiers

1540-36-9
156.225
MBXOOYPCIDHXGH-UHFFFAOYSA-N
73763
C9H16O2
MFCD00026235
3-Acetyl-2-heptanone
SDS Documents