Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-n-Butoxypropionitrile, 97%

$124.92 - $124.92

Chemical Identifiers

Chemical Name or Material 3-n-Butoxypropionitrile
CAS 6959-71-3
Molecular Formula CH3(CH2)3OCH2CH2CN
Formula Weight 127.19
MDL Number MFCD00045643
Beilstein 1747904
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1134214

  SDS
alfa aesar™
L1134214
25g Each for $124.92
Description & Specifications

Specifications

Formula Weight 127.19
Chemical Name or Material 3-n-Butoxypropionitrile
TSCA TSCA
Assay 97%
Molecular Formula CH3(CH2)3OCH2CH2CN
MDL Number MFCD00045643
CAS 6959-71-3
Boiling Point 105° to 106°C (15mmHg)
Beilstein 1747904
Quantity 25g
Flash Point 89°C (192°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UG1225000
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.