missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-n-Butoxypropionitrile, 97%

Manufacturer:  Alfa Aesar™ L1134206

 View more versions of this product

Catalog No. AAL1134206



Specifications

Specifications

3-n-Butoxypropionitrile
105°C to 106°C (15mmHg)
6959-71-3
MFCD00045643
3-butoxypropionitrile, propanenitrile, 3-butoxy, beta-butoxy propionitrile, propionitrile, 3-butoxy, 3-butoxypropannitril, 3-butoxypropiononitrile, 3-n-butoxypropionitrile, 3-butoxypropannitril czech, .beta.-butoxy propionitrile, 3-butoxypropanitrile
CCCCOCCC#N
127.187
127.19
89°C (192°F)
5g
C7H13NO
1747904
AWVNJBFNHGQUQU-UHFFFAOYSA-N
3-butoxypropanenitrile
23395
97%

Chemical Identifiers

6959-71-3
127.187
AWVNJBFNHGQUQU-UHFFFAOYSA-N
23395
CCCCOCCC#N
C7H13NO
MFCD00045643
3-butoxypropionitrile, propanenitrile, 3-butoxy, beta-butoxy propionitrile, propionitrile, 3-butoxy, 3-butoxypropannitril, 3-butoxypropiononitrile, 3-n-butoxypropionitrile, 3-butoxypropannitril czech, .beta.-butoxy propionitrile, 3-butoxypropanitrile
3-butoxypropanenitrile
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UG1225000

TSCA : TSCA

Documents