missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(N-BOC)-(2-aminoethyl)azetidine, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC451240010

 View more versions of this product

Catalog No. AC451240010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-(N-BOC)-(2-aminoethyl)azetidine
84°C to 86°C
162696-31-3
C10H20N2O2
CSPJUZSNJFANNU-UHFFFAOYSA-N
tert-butyl N-[2-(azetidin-3-yl)ethyl]carbamate
37818431
96%
Glass Bottle
1g
95% min. (HPLC)
2-azetin-3-yl-1-n-boc ethylamine, 3-2-n-boc-aminoethyl azetidine, 3-boc-aminoethylazetidine, 3-n-boc-aminoethyl azetidine, carbamic acid, n-2-3-azetidinyl ethyl-, 1,1-dimethylethyl ester, tert-butyl 2-3-azetidinyl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethyl carbamate, tert-butyl 2-azetidin-3-yl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethylcarbamate hydrochloride, tert-butyl n-2-azetidin-3-yl ethyl carbamate
CC(C)(C)OC(=O)NCCC1CNC1
200.28
200.28

Chemical Identifiers

162696-31-3
200.28
2-azetin-3-yl-1-n-boc ethylamine, 3-2-n-boc-aminoethyl azetidine, 3-boc-aminoethylazetidine, 3-n-boc-aminoethyl azetidine, carbamic acid, n-2-3-azetidinyl ethyl-, 1,1-dimethylethyl ester, tert-butyl 2-3-azetidinyl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethyl carbamate, tert-butyl 2-azetidin-3-yl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethylcarbamate hydrochloride, tert-butyl n-2-azetidin-3-yl ethyl carbamate
tert-butyl N-[2-(azetidin-3-yl)ethyl]carbamate
C10H20N2O2
CSPJUZSNJFANNU-UHFFFAOYSA-N
37818431
CC(C)(C)OC(=O)NCCC1CNC1
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

SDS Documents