missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(N-BOC)-(2-aminoethyl)azetidine, 96%, ACROS Organics™

Chemical Identifiers

CAS 162696-31-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
InChI Key CSPJUZSNJFANNU-UHFFFAOYSA-N
Synonym 2-azetin-3-yl-1-n-boc ethylamine, 3-2-n-boc-aminoethyl azetidine, 3-boc-aminoethylazetidine, 3-n-boc-aminoethyl azetidine, carbamic acid, n-2-3-azetidinyl ethyl-, 1,1-dimethylethyl ester, tert-butyl 2-3-azetidinyl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethyl carbamate, tert-butyl 2-azetidin-3-yl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethylcarbamate hydrochloride, tert-butyl n-2-azetidin-3-yl ethyl carbamate
PubChem CID 37818431
IUPAC Name tert-butyl N-[2-(azetidin-3-yl)ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCCC1CNC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC451242500
SDS View Documents
Thermo Fisher Scientific
AC451242500
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC451240010
SDS View Documents
Thermo Fisher Scientific
AC451240010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

162696-31-3
200.28
2-azetin-3-yl-1-n-boc ethylamine, 3-2-n-boc-aminoethyl azetidine, 3-boc-aminoethylazetidine, 3-n-boc-aminoethyl azetidine, carbamic acid, n-2-3-azetidinyl ethyl-, 1,1-dimethylethyl ester, tert-butyl 2-3-azetidinyl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethyl carbamate, tert-butyl 2-azetidin-3-yl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethylcarbamate hydrochloride, tert-butyl n-2-azetidin-3-yl ethyl carbamate
tert-butyl N-[2-(azetidin-3-yl)ethyl]carbamate
C10H20N2O2
CSPJUZSNJFANNU-UHFFFAOYSA-N
37818431
CC(C)(C)OC(=O)NCCC1CNC1

Specifications

84°C to 86°C
162696-31-3
C10H20N2O2
CSPJUZSNJFANNU-UHFFFAOYSA-N
tert-butyl N-[2-(azetidin-3-yl)ethyl]carbamate
37818431
96%
250mg
95% min. (HPLC)
2-azetin-3-yl-1-n-boc ethylamine, 3-2-n-boc-aminoethyl azetidine, 3-boc-aminoethylazetidine, 3-n-boc-aminoethyl azetidine, carbamic acid, n-2-3-azetidinyl ethyl-, 1,1-dimethylethyl ester, tert-butyl 2-3-azetidinyl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethyl carbamate, tert-butyl 2-azetidin-3-yl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethylcarbamate hydrochloride, tert-butyl n-2-azetidin-3-yl ethyl carbamate
CC(C)(C)OC(=O)NCCC1CNC1
200.28
200.28
3-(N-BOC)-(2-aminoethyl)azetidine
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

SDS