Please login to your online account to display your discounted pricing

3-MPPI, Tocris


α1 ligand

Quantity 50mg

Manufacturer: r & d systems  058150

 View more versions of this product

Catalog No. 058150

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 419.48
Chemical Name or Material 3-MPPI
Assay Percent Range >98%
Molecular Formula C23H25N5O3
CAS 133399-65-2
Synonym 2-[(4-Phenylpiperazin-1-yl)methyl]-2, 3-dihydroimidazo[1, 2-c]quinazolin-5(6H)-one
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material 3-MPPI
Assay Percent Range >98%
Molecular Formula C23H25N5O3
Formula Weight 419.48
Synonym 2-[(4-Phenylpiperazin-1-yl)methyl]-2, 3-dihydroimidazo[1, 2-c]quinazolin-5(6H)-one
Accessories

Store at Room Temperature