missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Morpholinopropanesulfonic Acid 98.0+%, TCI America™

$29.63 - $134.97

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-morpholinopropane-1-sulfonic acid, 3-morpholinopropanesulfonic acid, 3-n-morpholino propanesulfonic acid, 3-n-morpholinopropanesulfonic acid, 4-morpholinepropane sulfonic acid, 4-morpholinepropanesulfonic acid, mops, morpholinopropane sulfonic acid, morpholinopropanesulfonic acid, unii-273bp63nv3
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES C1COCCN1CCCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M070725G View Documents Tci America
M070725G
25g Each for $29.63
 
M0707250G View Documents Tci America
M0707250G
250g Each for $134.97
 
Specifications

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
3-morpholin-4-ylpropane-1-sulfonic acid
C7H15NO4S
MFCD00006183
3-morpholinopropane-1-sulfonic acid, 3-morpholinopropanesulfonic acid, 3-n-morpholino propanesulfonic acid, 3-n-morpholinopropanesulfonic acid, 4-morpholinepropane sulfonic acid, 4-morpholinepropanesulfonic acid, mops, morpholinopropane sulfonic acid, morpholinopropanesulfonic acid, unii-273bp63nv3
CHEBI:44115
C1COCCN1CCCS(=O)(=O)O

Specifications

White
1132-61-2
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
209.26
≥98.0% (T)
25g
C7H15NO4S
3-morpholinopropane-1-sulfonic acid, 3-morpholinopropanesulfonic acid, 3-n-morpholino propanesulfonic acid, 3-n-morpholinopropanesulfonic acid, 4-morpholinepropane sulfonic acid, 4-morpholinepropanesulfonic acid, mops, morpholinopropane sulfonic acid, morpholinopropanesulfonic acid, unii-273bp63nv3
C1COCCN1CCCS(=O)(=O)O
209.26
CHEBI:44115
Crystal-Powder at 20°C
3-Morpholinopropanesulfonic Acid [Good′s buffer component for biological research]
Safety and Handling

Safety and Handling

TSCA : Yes