missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-Morpholinophenyl)methylamine, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC33813DA

 View more versions of this product

Catalog No. CC33813DA


 

Specifications Description & Specifications

Specifications

(3-Morpholinophenyl)methylamine
96.5
Technical Grade
97%
192.26
Amber Glass Bottle
625470-29-3
100.0
97%
C11H16N2O
MFCD06659079
1g

Chemical Identifiers

625470-29-3
100.0
C11H16N2O
MFCD06659079
97%
96.5
97%
192.26
(3-Morpholinophenyl)methylamine
Technical Grade

Safety and Handling

SDS & Safety Documents

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger