Please login to your online account to display your discounted pricing

(3-Morpholinophenyl)methylamine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC33813CB

 View more versions of this product

Catalog No. CC33813CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 192.26
Purity Grade Technical Grade
Quantity 250mg
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H16N2O
MDL Number MFCD06659079
Chemical Name or Material (3-Morpholinophenyl)methylamine
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
CAS Min % 96.5
Packaging Amber Glass Bottle
Health Hazard 1 Danger
CAS 625470-29-3

Specifications

Chemical Name or Material (3-Morpholinophenyl)methylamine
CAS 625470-29-3
CAS Min % 96.5
CAS Max % 100.0
Purity Grade Technical Grade
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H16N2O
Formula Weight 192.26
MDL Number MFCD06659079
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

SDS & Safety Documents