missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-Morpholinophenyl)methylamine, 97%, Maybridge

$393.90 - $393.90

Chemical Identifiers

CAS 625470-29-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H16N2O
Formula Weight 192.26
MDL Number MFCD06659079
Chemical Name or Material (3-Morpholinophenyl)methylamine
Assay 97%
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC33813CB SDS Fisher Scientific Company
CC33813CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC33813DA SDS Fisher Scientific Company
CC33813DA
Amber Glass Bottle 1g Each for $393.90
Specifications Description & Specifications

Chemical Identifiers

625470-29-3
100.0
C11H16N2O
MFCD06659079
97%
96.5
97%
192.26
(3-Morpholinophenyl)methylamine
Technical Grade

Specifications

192.26
100.0
C11H16N2O
96.5
625470-29-3
97%
250mg
97%
MFCD06659079
Amber Glass Bottle
(3-Morpholinophenyl)methylamine
Technical Grade

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger