Please login to your online account to display your discounted pricing

3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate
CAS 137605-80-2
CAS Min % 96.5
CAS Max % 100.0
Molecular Formula HCl·H2O
Formula Weight 275.74
MDL Number MFCD09064949
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

CC40151CB

  SDS
fisher scientific company
CC40151CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC40151DA

  SDS
fisher scientific company
CC40151DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 275.74
CAS Min % 96.5
Health Hazard 1 Warning
CAS 137605-80-2
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Molecular Formula HCl·H2O
Chemical Name or Material 3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate
Assay 97%
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing
MDL Number MFCD09064949
Assay Percent Range 97%
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.