Please login to your online account to display your discounted pricing

3-(Morpholin-4-ylmethyl)aniline, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(Morpholin-4-ylmethyl)aniline
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H16N2O
Formula Weight 192.26
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC40114DE

  SDS
fisher scientific company
CC40114DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC40114CB

  SDS
fisher scientific company
CC40114CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC40114DA

  SDS
fisher scientific company
CC40114DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 192.26
Quantity 5g
Assay 95%
Chemical Name or Material 3-(Morpholin-4-ylmethyl)aniline
Molecular Formula C11H16N2O
Assay Percent Range 95%
CAS 123207-48-7
Packaging Amber Glass Bottle