Please login to your online account to display your discounted pricing

3-(Morpholin-4-ylmethyl)aniline, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC40114DA

 View more versions of this product

Catalog No. CC40114DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 192.26
Quantity 1g
Assay 95%
Assay Percent Range 95%
Chemical Name or Material 3-(Morpholin-4-ylmethyl)aniline
CAS 123207-48-7
Packaging Amber Glass Bottle
Molecular Formula C11H16N2O

Specifications

Chemical Name or Material 3-(Morpholin-4-ylmethyl)aniline
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H16N2O
Formula Weight 192.26