Please login to your online account to display your discounted pricing

3-morpholin-4-ylaniline, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC33814DE

 View more versions of this product

Catalog No. CC33814DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 178.23
Quantity 5g
Assay 97%
Chemical Name or Material 3-Morpholin-4-ylaniline
Assay Percent Range 97%
Molecular Formula C10H14N2O
CAS 159724-40-0
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 3-Morpholin-4-ylaniline
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H14N2O
Formula Weight 178.23