missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-morpholin-4-ylaniline, 97%, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C10H14N2O
Formula Weight 178.23
Chemical Name or Material 3-Morpholin-4-ylaniline
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC33814DE SDS Fisher Scientific Company
CC33814DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC33814DA SDS Fisher Scientific Company
CC33814DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC33814CB SDS Fisher Scientific Company
CC33814CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
178.23
97%
C10H14N2O
3-Morpholin-4-ylaniline

Specifications

159724-40-0
C10H14N2O
Amber Glass Bottle
3-Morpholin-4-ylaniline
97%
178.23
5g
97%