Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylvaleryl chloride, 97%

Chemical Identifiers

Chemical Name or Material 3-Methylvaleryl chloride
CAS 51116-72-4
Molecular Formula C6H11ClO
Formula Weight 134.61
MDL Number MFCD00039315
UN Number UN2920
Synonym 3-Methylpentanoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5041503

  SDS
alfa aesar™
H5041503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50415MD

  SDS
alfa aesar™
H50415MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.61
Synonym 3-Methylpentanoyl chloride
UN Number UN2920
Quantity 1g
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay 97%
Chemical Name or Material 3-Methylvaleryl chloride
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
MDL Number MFCD00039315
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 1 Danger
CAS 51116-72-4
Molecular Formula C6H11ClO

Applications
3-Methylvaleryl chloride can react with methylbenzene to product 4-methyl-1-p-tolyl-pentan-1-one. This reaction needs reagent AlCl3.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.